missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Disposable PD-10 Desalting Columns
Use for desalting and buffer exchange, as well as removal of low-molecular weight compounds. Cytiva Life Sciences™ Disposable PD-10 Desalting Columns provide fast, high-recovery.
Supplier: Cytiva 17085101
Description
- Packed with Sephadex™ G-25 medium
- Designed for ease of use and equilibration
- Prepare samples in range of 1.0 to 2.5mL by gravity flow or 1.75 to 2.5mL by centrifugation
- Desalt multiple samples in parallel with desalting capacity >90%
- Sample recovery in range of 70 to >95%
- Includes adapters for centrifugation
Specifications
Desalting Column, Sephadex G-25 Medium Packing | |
0.59 in. | |
8.3 mL | |
1.5 cm |
>50 μm (dry), 38 to 235 μm (wet) | |
Designed for ease of use and equilibration | |
4°C to 30°C | |
30 Pk. |