missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Dissection Tray

Aluminum dissection tray with wax liner.

Supplier:  Eisco™ BI0186QWX

Catalog No. S14000

Explore available promotions

Description

Description

  • 1 in. thick wax can be melted for easy renewal of pinning surface.
  • 35 x 24 x 5 cm (14 x 9 x 2 in.) approximately.
  • Works great with premium dissection pins 1 in. thick wax liner can be easily melted for resurfacing.
  • High quality aluminum construction.
Specifications

Specifications

Dissection Tray
Silver Tray, White Wax
Reusable
14 in.
10 in.
2 in.
Non-sterile
Not Autoclavable
Aluminum
14 x 10 x 2 in. (35 x 24 x 5 cm)
35 cm
24 cm
5 cm
Videos
SDS
Documents

Documents

Promotions

Promotions