Learn More
Dithizone, ACS, 85+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03328309
Description
Dithizone is used as an indicator for minute amounts of lead. It acts as a chelating agent for heavy metal poisoning. As a ligand, it forms complexes with metals like zinc, lead and mercury. Further, it is used to assess the purity of human pancreatic islet preparations, which is used for transplantation in patients with type 1 diabetes. In addition to this, it is used as a titrant in the quantitative determination of heavy metals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dithizone | |
168°C (decomposition) | |
C13H12N4S | |
10 g | |
14,3377 | |
Soluble in ethanol,chloroform,dimethyl sulfoxide and pyridine. Insoluble in water. | |
S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1 | |
256.33 | |
256.33 | |
ACS |
60-10-6 | |
Odorless | |
MFCD00003025 | |
748838 | |
dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone | |
UOFGSWVZMUXXIY-UHFFFAOYSA-N | |
1-anilino-3-phenyliminothiourea | |
657262 | |
≥85% | |
Crystalline |
Chemical Identifiers
60-10-6 | |
256.33 | |
UOFGSWVZMUXXIY-UHFFFAOYSA-N | |
657262 | |
S=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1 |
C13H12N4S | |
MFCD00003025 | |
dithizone, dithizon, ditizon, carbazone, diphenylthio, usaf ek-3092, diazenecarbothioic acid, phenyl-, 2-phenylhydrazide, 1,5-diphenyl-3-mercaptoformazan, 3-formazanthiol, 1,5-diphenyl, unii-njz2cj4d6p, 1,5-diphenyl-3-thiocarbazone | |
1-anilino-3-phenyliminothiourea |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-454-1
RTECSNumber : LQ9450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only