missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diuron, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S1760

Catalog No. S-1760


Explore available promotions

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Diuron330-54-11000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Diuron
2mL amber prescored glass ampule
0.1,99.9
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Methanol
CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl
233.092
CHEBI:116509
330-54-1,67-56-1
1 mL
0.1,99.9
Certified Reference Material
C9H10Cl2N2O
XMTQQYYKAHVGBJ-UHFFFAOYSA-N
3-(3,4-dichlorophenyl)-1,1-dimethylurea
3120

Chemical Identifiers

330-54-1
233.092
3120
3-(3,4-dichlorophenyl)-1,1-dimethylurea
C9H10Cl2N2O
XMTQQYYKAHVGBJ-UHFFFAOYSA-N
CHEBI:116509
CN(C)C(=O)NC1=CC(=C(C=C1)Cl)Cl
Videos
Safety and Handling

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only