missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Divinyl sulfone, 97%, stab. with 0.05% hydroquinone, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1282709

 View more versions of this product

Catalog No. AAL1282709

Explore available promotions

Description

Description

Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents, alkali metals and strong bases.
Specifications

Specifications

Divinyl sulfone
77-77-0
1.177
102°C (215°F)
1.476
10 g
1071329
Miscible with water.
C=CS(=O)(=O)C=C
118.15
CHEBI:53729
97%
stabilized with 0.05% hydroquinone
∼-26°C
233°C to 234°C
C4H6O2S
MFCD00008623
UN2927
divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
AFOSIXZFDONLBT-UHFFFAOYSA-N
1-ethenylsulfonylethene
6496
118.16

Chemical Identifiers

77-77-0
118.15
AFOSIXZFDONLBT-UHFFFAOYSA-N
6496
1-ethenylsulfonylethene
C4H6O2S
MFCD00008623
divinyl sulfone, vinyl sulfone, divinyl sulphone, bis ethenyl sulfone, vinyl sulphone, sulfone, divinyl, ethenesulfonyl-ethene, ethene, 1,1'-sulfonylbis, 1,1'-sulphonylbisethene, ethenesulfonyl ethene
CHEBI:53729
C=CS(=O)(=O)C=C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H310-H314-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 201-057-6

RTECSNumber : KM7175000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only