missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Divinylbenzene (m- and p- mixture) (contains Ethylvinylbenzene, Diethylbenzene) (stabilized with TBC) 50.0+%, TCI America™

Supplier:  TCI America D0958500ML

 View more versions of this product

Catalog No. D0958500ML


Explore available promotions

Description

Description

Specifications

Specifications

Divinylbenzene (m- and p- mixture) (contains Ethylvinylbenzene, Diethylbenzene) (stabilized with TBC
Yellow
C10H10
500 mL
divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
C=CC1=CC=CC=C1C=C
130.19
130.19
Liquid
1321-74-0
195°C
MFCD01778656,MFCD00010654
3082
MYRTYDVEIRVNKP-UHFFFAOYSA-N
1,2-diethenylbenzene
66666
≥50.0% (GC)

Chemical Identifiers

1321-74-0
130.19
MYRTYDVEIRVNKP-UHFFFAOYSA-N
66666
C=CC1=CC=CC=C1C=C
C10H10
MFCD01778656,MFCD00010654
divinylbenzene, 1,2-divinylbenzene, o-divinylbenzene, benzene, diethenyl, benzene, 1,2-diethenyl, divinylbenzen, divinyl benzen, divinyl-benzene, poly divinylbenzene, divinylbenzene dvb
1,2-diethenylbenzene
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0014

RTECSNumber : CZ9370000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions