missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-2-Amino-4-pentenoic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 174220050

 View more versions of this product

Catalog No. AC174220050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

DL-2-Amino-4-pentenoic acid
251°C to 253°C
Authentic
Glass bottle
H2C=CHCH2CH(NH2)CO2H
5 g
dl-allylglycine, 2-amino-4-pentenoic acid, dl-2-allylglycine, dl-2-amino-4-pentenoic acid, dl-c-allylglycine, 4-pentenoic acid, 2-amino, dl-2-amino-1-pentenoic acid, dl-2-aminopent-4-enoic acid, s-allyl-glycine, alpha-allyl-gly-oh
C=CCC(C(=O)O)N
115.13
115.13
Crystalline Powder
7685-44-1
White
99%
C5H9NO2
MFCD00063103
04, 467
WNNNWFKQCKFSDK-UHFFFAOYSA-N
2-aminopent-4-enoic acid
14044
99%

Chemical Identifiers

7685-44-1
115.13
WNNNWFKQCKFSDK-UHFFFAOYSA-N
14044
C=CCC(C(=O)O)N
C5H9NO2
MFCD00063103
dl-allylglycine, 2-amino-4-pentenoic acid, dl-2-allylglycine, dl-2-amino-4-pentenoic acid, dl-c-allylglycine, 4-pentenoic acid, 2-amino, dl-2-amino-1-pentenoic acid, dl-2-aminopent-4-enoic acid, s-allyl-glycine, alpha-allyl-gly-oh
2-aminopent-4-enoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-689-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only