missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-alpha-Lipoic Acid 99.0+%, TCI America™

Supplier:  TCI America L005825G

 View more versions of this product

Catalog No. L005825G


Explore available promotions

Description

Description

Specifications

Specifications

DL-alpha-Lipoic Acid
Yellow
C8H14O2S2
25 g
AGBQKNBQESQNJD-UHFFFAOYNA-N
5-(1,2-dithiolan-3-yl)pentanoic acid
864
206.32
Crystalline Powder
1077-28-7
165°C
MFCD00005474
dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
OC(=O)CCCCC1CCSS1
206.32
CHEBI:16494
≥99.0% (T)

Chemical Identifiers

1077-28-7
206.32
AGBQKNBQESQNJD-UHFFFAOYNA-N
864
5-(1,2-dithiolan-3-yl)pentanoic acid
C8H14O2S2
MFCD00005474
dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
CHEBI:16494
OC(=O)CCCCC1CCSS1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0817&(5)-0535

RTECSNumber : JP1192000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions