missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-alpha-Tocopherol, 97+%, Thermo Scientific Chemicals

$108.81 - $1363.37

Chemical Identifiers

CAS 10191-41-0
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.72
MDL Number MFCD00072051
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
PubChem CID 14985
ChEBI CHEBI:18145
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1703918
View Documents
Thermo Scientific Chemicals
A1703918
50 g
Each for $108.81
 
AAA1703930
View Documents
Thermo Scientific Chemicals
A1703930
250 g
Each for $389.37
 
AAA170390B
View Documents
Thermo Scientific Chemicals
A170390B
1000 g
Each for $1,363.37
 
Description

Description

DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-α-Tocopherol is used as an antioxidant that protects cell membrane lipids from oxidative damage.

Solubility
Miscible with chloroform, vegetable oils,  ether, acetone and alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible withstrong oxidizing agents.
Specifications

Chemical Identifiers

10191-41-0
430.72
GVJHHUAWPYXKBD-IEOSBIPESA-N
14985
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
C29H50O2
MFCD00072051
vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
CHEBI:18145
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1

Specifications

10191-41-0
0.950 g/mL
240°C (464°F)
C29H50O2
MFCD00072051
94012
14,10021
Miscible with chloroform,vegetable oils, ether,acetone and alcohol. Immiscible with water.
CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=C(C)C(O)=C(C)C(C)=C2O1
430.72
CHEBI:18145
≥97%
2°C to 4°C
200°C to 220°C (0.1 mm)
>97%
1.505
50 g
Air and light sensitive
vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
GVJHHUAWPYXKBD-IEOSBIPESA-N
(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
14985
430.72
DL-α-Tocopherol
Videos
Safety and Handling

Safety and Handling

P261-P272-P280g-P302+P352-P333+P313-P363-P501c

H317

EINECSNumber : 233-466-0

RTECSNumber : GA8746000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only