missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-alpha-Tocopherol Acetate 97.0+%, TCI America™
$55.37 - $465.40
Chemical Identifiers
CAS | 7695-91-2 |
---|---|
Molecular Formula | C31H52O3 |
Molecular Weight (g/mol) | 472.754 |
MDL Number | MFCD00072042 |
InChI Key | ZAKOWWREFLAJOT-CEFNRUSXSA-N |
Synonym | vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ |
PubChem CID | 86472 |
ChEBI | CHEBI:32321 |
IUPAC Name | [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate |
SMILES | CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C |
Chemical Identifiers
7695-91-2 | |
472.754 | |
ZAKOWWREFLAJOT-CEFNRUSXSA-N | |
86472 | |
[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate |
C31H52O3 | |
MFCD00072042 | |
vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ | |
CHEBI:32321 | |
CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C |
Specifications
7695-91-2 | |
224°C | |
MFCD00072042 | |
vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ | |
CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C | |
472.754 | |
CHEBI:32321 | |
≥98.0% (GC) | |
DL-alpha-Tocopherol Acetate |
Yellow | |
C31H52O3 | |
25 g | |
ZAKOWWREFLAJOT-CEFNRUSXSA-N | |
[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate | |
86472 | |
472.75 | |
Liquid |
Safety and Handling
EINECSNumber : (9)-0487
RTECSNumber : GA8747000
TSCA : Yes