missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Aminoglutethimide 98.0+%, TCI America™

Supplier:  TCI America A194725G

 View more versions of this product

Catalog No. A194725G


Explore available promotions

Description

Description

Specifications

Specifications

DL-Aminoglutethimide
152°C
C13H16N2O2
25 g
ROBVIMPUHSLWNV-UHFFFAOYSA-N
3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
2145
232.28
Crystalline Powder
125-84-8
White-Yellow
MFCD00010122
aminoglutethimide, dl-aminoglutethimide, cytadren, orimeten, p-aminoglutethimide, elipten, 3-4-aminophenyl-3-ethylpiperidine-2,6-dione, aminoglutetimida, 3-4-aminophenyl-3-ethyl-2,6-piperidinedione, aminoglutethimidum
CCC1(CCC(=O)NC1=O)C2=CC=C(C=C2)N
232.283
CHEBI:2654
≥98.0% (HPLC,T)

Chemical Identifiers

125-84-8
232.283
ROBVIMPUHSLWNV-UHFFFAOYSA-N
2145
3-(4-aminophenyl)-3-ethylpiperidine-2,6-dione
C13H16N2O2
MFCD00010122
aminoglutethimide, dl-aminoglutethimide, cytadren, orimeten, p-aminoglutethimide, elipten, 3-4-aminophenyl-3-ethylpiperidine-2,6-dione, aminoglutetimida, 3-4-aminophenyl-3-ethyl-2,6-piperidinedione, aminoglutethimidum
CHEBI:2654
CCC1(CCC(=O)NC1=O)C2=CC=C(C=C2)N
Videos
Safety and Handling

Safety and Handling

RTECSNumber : MA4026950

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions