Learn More
DL-Camphor, 96%
CAS: 76-22-2 | C10H16O | 152.24 g/mol
$73.32 - $452.79
Chemical Identifiers
| CAS | 76-22-2 |
|---|---|
| Molecular Formula | C10H16O |
| Molecular Weight (g/mol) | 152.24 |
| MDL Number | MFCD00074738,MFCD00064149 |
| InChI Key | DSSYKIVIOFKYAU-UHFFFAOYNA-N |
| Synonym | camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon |
| PubChem CID | 2537 |
| ChEBI | CHEBI:36773 |
| IUPAC Name | 4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
| SMILES | CC1(C)C2CCC1(C)C(=O)C2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC164642500
|
Thermo Scientific Chemicals
164642500 |
250 g | Plastic bottle |
Each for $73.32
|
|
||||
|
AC164640010
|
Thermo Scientific Chemicals
164640010 |
1 kg | Plastic bottle |
Each for $163.65
|
|
||||
|
AC164640051
|
Thermo Scientific Chemicals
164640051 |
5 kg | Plastic drum |
Each for $452.79
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 76-22-2 | |
| 152.24 | |
| DSSYKIVIOFKYAU-UHFFFAOYNA-N | |
| 2537 | |
| 4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
| C10H16O | |
| MFCD00074738,MFCD00064149 | |
| camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon | |
| CHEBI:36773 | |
| CC1(C)C2CCC1(C)C(=O)C2 |
Specifications
| 76-22-2 | |
| 64g/mL | |
| 64°C | |
| Plastic bottle | |
| MFCD00074738,MFCD00064149 | |
| 07,IV,213 | |
| camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon | |
| DSSYKIVIOFKYAU-UHFFFAOYNA-N | |
| 4,7,7-trimethylbicyclo[2.2.1]heptan-3-one | |
| 2537 | |
| 152.24 | |
| DL-Camphor |
| 175°C to 177°C | |
| 204°C | |
| 95% min. (GC) | |
| C10H16O | |
| 250 g | |
| 15,1734 | |
| Solubility in water: 1.5g/L (20°C). Other solubilities: at 25°C 1 g dissolves in 1 mL alcohol,1 mL ether,,0.5 mL chloroform,0.4 mL benzene,0.4 mL acetone,,1.5 mL turpentine oil,0.5 mL glacial acetic acid.,soluble in aniline,nitrobenzene,c | |
| CC1(C)C2CCC1(C)C(=O)C2 | |
| 152.24 | |
| CHEBI:36773 | |
| 96% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable solid.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 200-945-
RUO – Research Use Only