missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Carnitine hydrochloride, 98+%, Thermo Scientific Chemicals

$46.84 - $139.79

Chemical Identifiers

CAS 461-05-2
Molecular Formula C7H17ClNO3+
Molecular Weight (g/mol) 198.667
MDL Number MFCD00011904
InChI Key JXXCENBLGFBQJM-UHFFFAOYSA-O
Synonym dl-carnitine hydrochloride, 3-carboxy-2-hydroxypropyl-trimethylammonium hydrochloride, 2,4-bis oxidanyl-4-oxidanylidene-butyl-trimethyl-azanium hydrochloride
PubChem CID 24206429
IUPAC Name (3-carboxy-2-hydroxypropyl)-trimethylazanium;hydrochloride
SMILES C[N+](C)(C)CC(CC(=O)O)O.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1658914
View Documents
Thermo Scientific Chemicals
A1658914
25 g
Each for $46.84
 
AAA1658922
View Documents
Thermo Scientific Chemicals
A1658922
100 g
Each for $139.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

461-05-2
198.667
JXXCENBLGFBQJM-UHFFFAOYSA-O
24206429
C[N+](C)(C)CC(CC(=O)O)O.Cl
C7H17ClNO3+
MFCD00011904
dl-carnitine hydrochloride, 3-carboxy-2-hydroxypropyl-trimethylammonium hydrochloride, 2,4-bis oxidanyl-4-oxidanylidene-butyl-trimethyl-azanium hydrochloride
(3-carboxy-2-hydroxypropyl)-trimethylazanium;hydrochloride

Specifications

461-05-2
C7H17ClNO3+
25 g
Hygroscopic
dl-carnitine hydrochloride, 3-carboxy-2-hydroxypropyl-trimethylammonium hydrochloride, 2,4-bis oxidanyl-4-oxidanylidene-butyl-trimethyl-azanium hydrochloride
C[N+](C)(C)CC(CC(=O)O)O.Cl
198.667
197.66
DL-Carnitine hydrochloride
196°C to 200°C
MFCD00011904
4163212
14,1849
JXXCENBLGFBQJM-UHFFFAOYSA-O
(3-carboxy-2-hydroxypropyl)-trimethylazanium;hydrochloride
24206429
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 207-309-1

RTECSNumber : BP2979140

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only