missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Cysteine Hydrochloride Monohydrate 98.0+%, TCI America™
Supplier: TCI America C051625G
Specifications
DL-Cysteine Hydrochloride Monohydrate | |
White | |
MFCD00151030 | |
QIJRTFXNRTXDIP-UHFFFAOYNA-N | |
2-amino-3-sulfanylpropanoic acid hydrate hydrochloride | |
12219647 | |
≥98.0% (T) | |
Crystalline Powder |
116797-51-4 | |
C3H10ClNO3S | |
H-DL-Cys-OH.HCl | |
O.Cl.NC(CS)C(O)=O | |
175.63 | |
157.61 | |
25 g |
Chemical Identifiers
116797-51-4 | |
175.63 | |
QIJRTFXNRTXDIP-UHFFFAOYNA-N | |
12219647 | |
O.Cl.NC(CS)C(O)=O |
C3H10ClNO3S | |
MFCD00151030 | |
H-DL-Cys-OH.HCl | |
2-amino-3-sulfanylpropanoic acid hydrate hydrochloride |
Safety and Handling
TSCA : No