missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Dithiothreitol, ≥99.0%, MP Biomedicals™
$100.97 - $1569.18
Chemical Identifiers
CAS | 27565-41-9 |
---|---|
Molecular Formula | C4H10O2S2 |
Molecular Weight (g/mol) | 154.24 |
MDL Number | MFCD00004877,MFCD00064305 |
InChI Key | VHJLVAABSRFDPM-UHFFFAOYNA-N |
Synonym | DTT, 1, 4-Dithio-DL-threitol, Cleland Reagent |
IUPAC Name | 1,4-disulfanylbutane-2,3-diol |
SMILES | OC(CS)C(O)CS |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
ICN10059701
|
MP Biomedicals Inc
0210059701 |
1 g |
Each for $100.97
|
|
|||||
ICN10059705
|
MP Biomedicals Inc
0210059705 |
5 g |
Each for $235.17
|
|
|||||
ICN10059710
|
MP Biomedicals Inc
0210059710 |
10 g |
Each for $466.63
|
|
|||||
ICN10059725
|
MP Biomedicals Inc
0210059725 |
25 g |
Each for $955.47
|
|
|||||
ICN10059750
|
MP Biomedicals Inc
0210059750 |
50 g |
Each for $1,569.18
|
|
|||||
Chemical Identifiers
27565-41-9 | |
154.24 | |
VHJLVAABSRFDPM-UHFFFAOYNA-N | |
1,4-disulfanylbutane-2,3-diol |
C4H10O2S2 | |
MFCD00004877,MFCD00064305 | |
DTT, 1, 4-Dithio-DL-threitol, Cleland Reagent | |
OC(CS)C(O)CS |
Specifications
27565-41-9 | |
≥99.0% | |
MFCD00004877,MFCD00064305 | |
DTT, 1, 4-Dithio-DL-threitol, Cleland Reagent | |
OC(CS)C(O)CS | |
154.24 | |
Powder |
White | |
C4H10O2S2 | |
1 g | |
VHJLVAABSRFDPM-UHFFFAOYNA-N | |
1,4-disulfanylbutane-2,3-diol | |
154.25 | |
DL-Dithiothreitol |
Safety and Handling
EINECSNumber : 248-531-9