missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Dithiothreitol, 99.5%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0219482125

 View more versions of this product

Catalog No. ICN19482125


Explore available promotions

Description

Description

Specifications

Specifications

DL-Dithiothreitol
>99%
MFCD00004877,MFCD00064305
Cleland's reagent, DTT
OC(CS)C(O)CS
154.24
Molecular Biology Reagent
27565-41-9
C4H10O2S2
25 g
VHJLVAABSRFDPM-UHFFFAOYNA-N
1,4-disulfanylbutane-2,3-diol
154.2
Powder

Chemical Identifiers

27565-41-9
154.24
VHJLVAABSRFDPM-UHFFFAOYNA-N
1,4-disulfanylbutane-2,3-diol
C4H10O2S2
MFCD00004877,MFCD00064305
Cleland's reagent, DTT
OC(CS)C(O)CS
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 248-531-9

Recommended Storage : Store at + 4°C

SDS
Documents

Documents

Promotions

Promotions