missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Glutamine, 98%, Thermo Scientific Chemicals
$82.92 - $316.93
Chemical Identifiers
CAS | 6899-04-3 |
---|---|
Molecular Formula | C5H10N2O3 |
Molecular Weight (g/mol) | 146.146 |
MDL Number | MFCD00065103 |
InChI Key | ZDXPYRJPNDTMRX-UHFFFAOYSA-N |
Synonym | dl-glutamine, glutamin, 2-amino-4-carbamoylbutanoic acid, dl glutamine, glutamine dl-form, glutamine, dl, l-2-aminoglutaramic acid, 2,5-bis azanyl-5-oxidanylidene-pentanoic acid, gamma-glutamine, .gamma.-glutamine |
PubChem CID | 738 |
ChEBI | CHEBI:28300 |
IUPAC Name | 2,5-diamino-5-oxopentanoic acid |
SMILES | C(CC(=O)N)C(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6291503
|
Thermo Scientific Chemicals
H6291503 |
1 g |
Each for $82.92
|
|
|||||
AAH6291506
|
Thermo Scientific Chemicals
H6291506 |
5 g |
Each for $316.93
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6899-04-3 | |
146.146 | |
ZDXPYRJPNDTMRX-UHFFFAOYSA-N | |
738 | |
2,5-diamino-5-oxopentanoic acid |
C5H10N2O3 | |
MFCD00065103 | |
dl-glutamine, glutamin, 2-amino-4-carbamoylbutanoic acid, dl glutamine, glutamine dl-form, glutamine, dl, l-2-aminoglutaramic acid, 2,5-bis azanyl-5-oxidanylidene-pentanoic acid, gamma-glutamine, .gamma.-glutamine | |
CHEBI:28300 | |
C(CC(=O)N)C(C(=O)O)N |
Specifications
177°C | |
C5H10N2O3 | |
14,4471 | |
ZDXPYRJPNDTMRX-UHFFFAOYSA-N | |
2,5-diamino-5-oxopentanoic acid | |
738 | |
146.14 | |
1 g |
6899-04-3 | |
MFCD00065103 | |
dl-glutamine, glutamin, 2-amino-4-carbamoylbutanoic acid, dl glutamine, glutamine dl-form, glutamine, dl, l-2-aminoglutaramic acid, 2,5-bis azanyl-5-oxidanylidene-pentanoic acid, gamma-glutamine, .gamma.-glutamine | |
C(CC(=O)N)C(C(=O)O)N | |
146.146 | |
CHEBI:28300 | |
98% | |
DL-Glutamine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only