missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DL-Isocitric acid, trisodium salt hydrate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 205010010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
DL-Isocitric acid, trisodium salt hydrate | |
White | |
95% | |
C6H5Na3O7 | |
MFCD00149284 | |
03, 555 | |
Solubility in water: soluble in water | |
[Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O | |
258.07 | |
258.07 | |
Crystalline Powder |
1637-73-6 | |
Authentic | |
Glass bottle | |
NaO2CCH2CH(CO2Na)CH(OH)CO2Na·xH2O | |
1 g | |
dl-isocitric acid, isocitric acid trisodium salt, sodium isocitrate, dl-isocitric acid trisodium salt hydrate, dl-isocitric acid trisodium salt, trisodium 1-hydroxypropane-1,2,3-tricarboxylate, trisodium isocitrate, tripotassium isocitrate, isocitric acid, dl, dl-isocitric acid, trisodium salt hydrate | |
HWMVXEKEEAIYGB-UHFFFAOYNA-K | |
trisodium;1-hydroxypropane-1,2,3-tricarboxylate | |
168942 | |
95% |
Chemical Identifiers
1637-73-6 | |
258.07 | |
HWMVXEKEEAIYGB-UHFFFAOYNA-K | |
168942 | |
[Na+].[Na+].[Na+].OC(C(CC([O-])=O)C([O-])=O)C([O-])=O |
C6H5Na3O7 | |
MFCD00149284 | |
dl-isocitric acid, isocitric acid trisodium salt, sodium isocitrate, dl-isocitric acid trisodium salt hydrate, dl-isocitric acid trisodium salt, trisodium 1-hydroxypropane-1,2,3-tricarboxylate, trisodium isocitrate, tripotassium isocitrate, isocitric acid, dl, dl-isocitric acid, trisodium salt hydrate | |
trisodium;1-hydroxypropane-1,2,3-tricarboxylate |
Safety and Handling
EINECSNumber : 216-668-3
RUO – Research Use Only