missing translation for 'onlineSavingsMsg'
Learn More

DL-Mandelic Acid 99.0+%, TCI America™

$55.92 - $149.13

Chemical Identifiers

CAS 90-64-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00064250
InChI Key IWYDHOAUDWTVEP-UHFFFAOYSA-N
Synonym mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid
PubChem CID 1292
ChEBI CHEBI:35825
IUPAC Name 2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M003825G
View Documents
TCI America
M003825G
25 g
Each for $55.92
Only null left
 
M0038500G
View Documents
TCI America
M0038500G
500 g
Each for $149.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

90-64-2
152.149
IWYDHOAUDWTVEP-UHFFFAOYSA-N
1292
2-hydroxy-2-phenylacetic acid
C8H8O3
MFCD00064250
mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid
CHEBI:35825
C1=CC=C(C=C1)C(C(=O)O)O

Specifications

121°C
White-Yellow
MFCD00064250
IWYDHOAUDWTVEP-UHFFFAOYSA-N
2-hydroxy-2-phenylacetic acid
1292
152.15
25 g
DL-Mandelic Acid
90-64-2
C8H8O3
mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid
C1=CC=C(C=C1)C(C(=O)O)O
152.149
CHEBI:35825
≥99.0% (GC,T)
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1764

RTECSNumber : OO6300000

TSCA : Yes

SDS
Documents

Documents