Learn More
DL-Menthol, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1809830
Description
Used in perfumery to give power, lift, and freshness. For use in toothpaste, cough drops, mouthwash, ice cream, chewing gum, tobacco, medical plaster, tonic, and shampoos. Because of its cooling action, it is to be found in a wide range of medicines treating sore throats and mouth irritations. It is also used in treatments for minor aches and sprains, and in nasal decongestants. It is found in some oral hygiene products to combat bad breath. Of course, it is present in a number of chewing gums. It was first added to cigarettes in the 1920s and 1930s (e.g. Spud and Kool brands) for both its flavor and also to reduce irritation, especially to the throat.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in alcohol, paraffin oil and water, 434.5 mg/L @ 25°C (est).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
DL-Menthol | |
33°C to 36°C | |
212°C to 216°C | |
Sweet | |
1.4615 | |
250 g | |
14,5837 | |
Soluble in alcohol,paraffin oil and water,434.5mg/L @ 25°C (est). | |
CC1CCC(C(C1)O)C(C)C | |
156.269 | |
156.27 |
89-78-1 | |
0.89 | |
93°C (199°F) | |
C10H20O | |
MFCD00001484 | |
3194263 | |
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol | |
NOOLISFMXDJSKH-IVZWLZJFSA-N | |
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol | |
6566020 | |
≥98% |
Chemical Identifiers
89-78-1 | |
156.269 | |
NOOLISFMXDJSKH-IVZWLZJFSA-N | |
6566020 | |
CC1CCC(C(C1)O)C(C)C |
C10H20O | |
MFCD00001484 | |
menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol | |
(1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-939-0
RTECSNumber : OT0350000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only