missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DL-Serine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

$55.88 - $821.78

Chemical Identifiers

CAS 5619-04-5
Molecular Formula C4H10ClNO3
Molecular Weight (g/mol) 155.58
MDL Number MFCD00012593
InChI Key NDBQJIBNNUJNHA-UHFFFAOYNA-N
Synonym dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908
PubChem CID 517373
IUPAC Name methyl 2-amino-3-hydroxypropanoate;hydrochloride
SMILES COC(=O)C(CO)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1390409
View Documents
Thermo Scientific Chemicals
A1390409
10 g N/A
 
AAA1390418
View Documents
Thermo Scientific Chemicals
A1390418
50 g N/A
 
AAA1390430
View Documents
Thermo Scientific Chemicals
A1390430
250 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5619-04-5
155.58
NDBQJIBNNUJNHA-UHFFFAOYNA-N
517373
COC(=O)C(CO)N.Cl
C4H10ClNO3
MFCD00012593
dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908
methyl 2-amino-3-hydroxypropanoate;hydrochloride

Specifications

∼134°C (decomposition)
C4H10ClNO3
6067970
NDBQJIBNNUJNHA-UHFFFAOYNA-N
methyl 2-amino-3-hydroxypropanoate;hydrochloride
155.58
155.58
≥98%
5619-04-5
MFCD00012593
dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908
COC(=O)C(CO)N.Cl
10 g
517373
Moisture sensitive
DL-Serine methyl ester hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-047-1

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only