missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals DL-Thioctic Acid, 98+%

Supplier:  Thermo Scientific Chemicals 138721000

 View more versions of this product

Catalog No. AC138721000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

DL-Thioctic acid
60°C to 63°C
160°C to 165°C
Authentic
Glass bottle
MFCD00005474
15, 9479
(2% in 10% NaOH) Clear yellow
OC(=O)CCCCC1CCSS1
206.32
CHEBI:16494
98+%
1077-28-7
Yellow
1% max.
98+%
C8H14O2S2
100 g
dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
AGBQKNBQESQNJD-UHFFFAOYNA-N
5-(dithiolan-3-yl)pentanoic acid
864
206.32
Crystalline Powder or Crystals

Chemical Identifiers

1077-28-7
206.32
AGBQKNBQESQNJD-UHFFFAOYNA-N
864
5-(dithiolan-3-yl)pentanoic acid
C8H14O2S2
MFCD00005474
dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
CHEBI:16494
OC(=O)CCCCC1CCSS1
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 214-071-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only