missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals DL-α-Tocopheryl acetate, 98%
$66.22 - $66.22
Chemical Identifiers
CAS | 7695-91-2 |
---|---|
Molecular Formula | C31H52O3 |
Molecular Weight (g/mol) | 472.75 |
MDL Number | MFCD00072042 |
InChI Key | ZAKOWWREFLAJOT-CEFNRUSXSA-N |
Synonym | vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ |
PubChem CID | 86472 |
ChEBI | CHEBI:32321 |
IUPAC Name | [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate |
SMILES | CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC421070010
|
Thermo Scientific Chemicals
421070010 |
1 g | Glass bottle |
Each for $66.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7695-91-2 | |
472.75 | |
ZAKOWWREFLAJOT-CEFNRUSXSA-N | |
86472 | |
[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate |
C31H52O3 | |
MFCD00072042 | |
vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ | |
CHEBI:32321 | |
CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C |
Specifications
7695-91-2 | |
257°C | |
97.5% min. (GC) | |
C31H52O3 | |
1 g | |
vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ | |
ZAKOWWREFLAJOT-CEFNRUSXSA-N | |
[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate | |
86472 | |
472.75 | |
Viscous Liquid |
Green to Yellow | |
Authentic | |
Glass bottle | |
MFCD00072042 | |
15, 9653 | |
Solubility in water: slightly soluble | |
CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C | |
472.75 | |
CHEBI:32321 | |
98% | |
DL-α-Tocopheryl acetate |
Safety and Handling
EINECSNumber : 231-710-
RUO – Research Use Only