Learn More
Leica Microsystems DM300 Monocular Life Sciences Educational Microscope
The Leica DM300 microscope is an educational tool designed for classrooms. With user-friendly design and reliable optics, it offers an accessible and engaging platform for hands-on learning, making microscopy an effective and enjoyable educational journey for students.
Supplier: Leica Microsystems 8122680
Description
The Leica DM300 monocular microscope is a purpose-built instrument designed to enrich educational experiences. Tailored for classroom settings, this microscope offers a straightforward and educational platform for students to explore the microscopic realm. With its user-friendly design and reliable optics, the Leica DM300 monocular version provides an accessible and engaging tool for educators dedicated to fostering hands-on learning. Elevate your classroom with this microscope, designed to make the study of microscopy an educational journey that is both effective and enjoyable for students.
- DM 300 Right Hand Stage Stand with 4 position nosepiece
- Universal Power Supply, mechanical stage with slide holder
- Pre-Centered and Pre-Focused Abbe condenser with adjustable aperture diaphragm and labels for matching objective magnifications
- LED illumination with continuous intensity control
- Dust cover
- User Documents
- 45 degree Monocular EZ Tube with integrated 10X/18 eyepiece with pointer and preset diopter
- Achromat Objectives 4X/0.10NA, 10X/0.25 NA, 40X/0.65 NA, 100x/1.25 NA
- Oil Immersion, 10ml oil
- 13613383 DM300 Binoc, Mech. Stage, Abbe, 4 Obj.
- 13613900 U.S. power cord
Specifications
10X/18 mm, with pointer | |
LED, Transmitted Light | |
DM300 | |
Educational Microscope |
Monocular | |
4X, 10X, 40X, 100X | |
Achromatic | |
100/240 V |