Learn More
Gibco™ DMEM, low glucose, GlutaMAX™ Supplement, pyruvate
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells.
Supplier: Gibco™ 10567022
Description
Dulbecco's Modified Eagle Medium (DMEM) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This DMEM is modified as follows:
- With: Low Glucose, Sodium Pyruvate, GlutaMAX™, Phenol Red
- Without: HEPES
The complete formulation is available.
DMEM is unique from other media as it contains 4 times the concentration of amino acids and vitamins than the original Eagle's Minimal Essential Medium. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. Gibco™ DMEM with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format.
DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM uses a sodium bicarbonate buffer system (3.7 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
Specifications
| HeLa, 293, Cos-7, and PC-12 | |
| Animal Origin-free | |
| Liquid | |
| Standard Serum Supplementation | |
| Sterile-filtered | |
| No HEPES | |
| GlutaMAX | |
| Room Temperature |
| Primary Fibroblasts, Neurons, Glial Cells, HUVECs, Smooth Muscle Cells | |
| 1X | |
| DMEM (Dulbecco's Modified Eagle Medium) | |
| Sterile-filtered | |
| GlutaMAX, Low Glucose, Sodium Pyruvate, Phenol Red | |
| cGMP-compliant under the ISO 13485 standard | |
| 10 x 500 mL | |
| Store at 2°C to 8°C and protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.