missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DMF, Dimethylformamide, GC Headspace Grade, Thermo Scientific™
DMF processed to high purity for accurate and repeatable determination of trace levels of residual solvents.
Supplier: Thermo Fisher Scientific D1331
Description
- Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
- 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
- This product is UV Absorbance Tested to ensure low organic contamination
- Processed for low water content to facilitate the extraction of organic volatile impurities, or VOC
Specifications
DMF, Dimethylformamide | |
0.300 AU Maximum (275nm), 0.050 AU Maximum (300nm), 0.005 AU Maximum (350nm), 0.005 AU Maximum (400nm) | |
1 L | |
Glass Bottle | |
5 ppm Maximum | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
≥99.9% |
68-12-2 | |
Colorless | |
Pass Test | |
Liquid | |
C3H7NO | |
0.03% Maximum | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09 | |
GC Headspace |
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Safety and Handling
Recommended Storage : Room temperature, packaged under inert atmosphere.