Learn More
Enzymax Llc DNA Mini spin column only, 100/pk DFS Item
Supplier:  Enzymax EZC101
These popular Mini DNA columns have been widely used as economic substitutes for all major manufacturers plasmid DNA, viral and genomic DNA Kits including Qiagen (Qiaprep, Qiaquick, QiaAmp, DNeasy), Sigma (GeneElute), Invitrogen (PureLink DNA), and Promega (PureYield DNA). We also provide solution recipes for DNA mini-prep, DNA gel extraction, and PCR clean up for users who want to make their own solutions.
Catalog No. NC1013737
Only null left
Explore available promotions