MP Biomedicals™ DNA-OFF™
DNA-OFF™ is a ready to use non-alkaline cleaning solution that is highly active against DNA contamination. It is non-carcinogenic, non corosive and heat resistant solution used for eliminating DNA from surfaces, such as PCR workstations, countertops, apparatus and pipettors.
Supplier: MP Biomedicals™ 11QD0500
Description
- The presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts.
- This ready-to-use solution is highly active against DNA contamination and is intended for use at PCR workstations, including countertops, apparatus and pipettors.
- It is stable, heat resistant, non-corrosive and free of carcinogenic compounds.
Specifications
Removal Agent | |
500 mL | |
Intended for use at PCR workstations, including countertops, apparatus and pipettors |
16.9 oz. | |
Highly active non-alkaline coleaning solution for DNA contamination, ready to use. | |
Bottle |