Thermo Scientific™ DNase/RNase-Free Deionized Water
Designed for use in all molecular biology applications.
Supplier:  Thermo Scientific™ LSGA57775
Catalog No. FERA57775
Explore available promotions
Description
Description
DNase/RNase-Free Distilled Water is designed for use in all molecular biology applications. It is 0.1-μm membrane-filtered and tested for DNase and RNase activity.
Specifications
Specifications
Nuclease-free deionized water
Molecular Biology
Not DEPC-Treated
1 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only.