missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dobutamine hydrochloride, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J61333MD

 View more versions of this product

Catalog No. AAJ61333MD


Explore available promotions

Description

Description

Dobutamine hydrochloride is a catecholamine that acts as a β-adrenergic receptor agonist. It has its strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions. As a result, it strongly increases cardiac contractility with modest chronotropic, arrhythmogenic, and vascular side effects. It is also a polyphenolic antioxidant and can inhibit all human carbonic anhydrase isoforms with Ki values near 1M.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dobutamine hydrochloride is a catecholamine that acts as a β-adrenergic receptor agonist. It has its strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions. As a result, it strongly increases cardiac contractility with modest chronotropic, arrhythmogenic, and vascular side effects. It is also a polyphenolic antioxidant and can inhibit all human carbonic anhydrase isoforms with Ki values near 1 μM.

Solubility
Soluble in water or ethanol with warming. Also soluble in methanol

Notes
Solutions oxidize readily at pH greater than 11. Protect solutions from light
Specifications

Specifications

Dobutamine hydrochloride
184°C to 186°C
C18H24ClNO3
250 mg
dobutamine hydrochloride, dobutamine hcl, dobutrex, inotrex, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, +--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +-
BQKADKWNRWCIJL-UHFFFAOYNA-N
hydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride
65324
337.85
49745-95-1
White
MFCD00153795
14,3395
Soluble in water or ethanol with warming; Also soluble in methanol
[H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1
337.84
CHEBI:4671
Powder

Chemical Identifiers

49745-95-1
337.84
BQKADKWNRWCIJL-UHFFFAOYNA-N
65324
hydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride
C18H24ClNO3
MFCD00153795
dobutamine hydrochloride, dobutamine hcl, dobutrex, inotrex, dl-dobutamine hydrochloride, dobutamina clorhidrato spanish, 4-2-4-4-hydroxyphenyl butan-2-ylamino ethyl benzene-1,2-diol hydrochloride, +--4-2-3-p-hydroxyphenyl-1-methylpropyl amino ethyl pyrocatechol hydrochloride, 1,2-benzenediol, 4-2-3-4-hydroxyphenyl-1-methylpropyl amino ethyl-, hydrochloride, +-
CHEBI:4671
[H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302-H312-H332
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c

H302+H312+H332-H361

EINECSNumber : 256-464-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only