missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dodecanedioic Dihydrazide 98.0+%, TCI America™
Supplier: TCI America D2342500G
Specifications
Dodecanedioic Dihydrazide | |
190°C | |
C12H26N4O2 | |
500 g | |
GRGBENNNGZARRZ-UHFFFAOYSA-N | |
dodecanedihydrazide | |
20013 | |
≥98.0% (T) |
4080-98-2 | |
White-Yellow | |
MFCD00043619 | |
Dodecanediohydrazide | |
C(CCCCCC(=O)NN)CCCCC(=O)NN | |
258.366 | |
258.37 | |
Crystalline Powder |
Chemical Identifiers
4080-98-2 | |
258.366 | |
GRGBENNNGZARRZ-UHFFFAOYSA-N | |
20013 | |
C(CCCCCC(=O)NN)CCCCC(=O)NN |
C12H26N4O2 | |
MFCD00043619 | |
Dodecanediohydrazide | |
dodecanedihydrazide |
Safety and Handling
EINECSNumber : (2)-3246
RTECSNumber : JR2400000
TSCA : Yes