missing translation for 'onlineSavingsMsg'
Learn More

Dodecyl Sulfide 96.0+%, TCI America™

$111.84 - $1084.87

Chemical Identifiers

CAS 2469-45-6
Molecular Formula C24H50S
Molecular Weight (g/mol) 370.724
MDL Number MFCD00008975
InChI Key UPYPTOCXMIWHSG-UHFFFAOYSA-N
Synonym dodecyl sulfide, didodecyl sulfide, dilauryl sulfide, n-dodecyl sulfide, dodecane, 1,1'-thiobis, bis dodecyl sulfide, bis n-dodecyl sulfide, unii-m8cf2nuw66, m8cf2nuw66, dodecylthiododecane
PubChem CID 72875
IUPAC Name 1-dodecylsulfanyldodecane
SMILES CCCCCCCCCCCCSCCCCCCCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D132725G
View Documents
TCI America
D132725G
25 g
Each for $111.84
Only null left
 
D1327500G
View Documents
TCI America
D1327500G
500 g
Each for $1,084.87
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2469-45-6
370.724
UPYPTOCXMIWHSG-UHFFFAOYSA-N
72875
CCCCCCCCCCCCSCCCCCCCCCCCC
C24H50S
MFCD00008975
dodecyl sulfide, didodecyl sulfide, dilauryl sulfide, n-dodecyl sulfide, dodecane, 1,1'-thiobis, bis dodecyl sulfide, bis n-dodecyl sulfide, unii-m8cf2nuw66, m8cf2nuw66, dodecylthiododecane
1-dodecylsulfanyldodecane

Specifications

2469-45-6
White-Yellow
MFCD00008975
dodecyl sulfide, didodecyl sulfide, dilauryl sulfide, n-dodecyl sulfide, dodecane, 1,1'-thiobis, bis dodecyl sulfide, bis n-dodecyl sulfide, unii-m8cf2nuw66, m8cf2nuw66, dodecylthiododecane
CCCCCCCCCCCCSCCCCCCCCCCCC
370.724
370.72
Crystalline Powder
41°C
C24H50S
25 g
UPYPTOCXMIWHSG-UHFFFAOYSA-N
1-dodecylsulfanyldodecane
72875
≥96.0% (GC)
Dodecyl Sulfide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0473&(2)-0475

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'