missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Dodecylamine | |
White | |
C12H27N | |
25 g | |
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 | |
CCCCCCCCCCCCN | |
185.36 | |
185.36 | |
Crystalline Powder |
124-22-1 | |
259°C | |
MFCD00008154 | |
3259 | |
JRBPAEWTRLWTQC-UHFFFAOYSA-N | |
dodecan-1-amine | |
13583 | |
≥97.0% (GC,T) |
Chemical Identifiers
124-22-1 | |
185.36 | |
JRBPAEWTRLWTQC-UHFFFAOYSA-N | |
13583 | |
CCCCCCCCCCCCN |
C12H27N | |
MFCD00008154 | |
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4 | |
dodecan-1-amine |
Safety and Handling
EINECSNumber : (2)-0133
RTECSNumber : JR6475000
TSCA : Yes