missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dodecyltrimethylammonium chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 409320250

 View more versions of this product

Catalog No. AC409320250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Dodecyltrimethylammonium chloride
237.0°C
Authentic
Glass bottle
CH3(CH2)11N(CH3)3Cl
25 g
(10% in water) Clear
CCCCCCCCCCCC[N+](C)(C)C.[Cl-]
263.88
263.88
Chunks and/or Powder
112-00-5
White to Yellow
97.5% min. (Argentometry)
C15H34ClN
MFCD00041974
dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
DDXLVDQZPFLQMZ-UHFFFAOYSA-M
dodecyl(trimethyl)azanium;chloride
8152
98%

Chemical Identifiers

112-00-5
263.88
DDXLVDQZPFLQMZ-UHFFFAOYSA-M
8152
CCCCCCCCCCCC[N+](C)(C)C.[Cl-]
C15H34ClN
MFCD00041974
dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
dodecyl(trimethyl)azanium;chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-927-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only