Thermo Scientific™ Domed and Flat PCR Cap Strips
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Supplier:  Thermo Scientific™ AB0786
 View more versions of this product
Catalog No. AB-0786
Explore available promotions
Description
Description
These PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
Natural
Virgin Medical Grade Polypropylene
300
25 packs of 12
Strips of 8 Flat Caps
Certified free of human DNA, RNase, DNase, and endotoxins.
96
12
Facilitates sample labeling
96-well PCR Plate, Standard and Low-profile PCR Tube Strips