Thermo Scientific™ Ultra Clear qPCR Caps, strips of 8
Optimize PCR applications with these cap strips for use with Thermo Scientific 96-well PCR plates and strip tubes.
Supplier:  Thermo Scientific™ AB0866
Catalog No. AB-0866
Explore available promotions
Description
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Highlights
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Specifications
Specifications
Clear
8
Strips of 8 Optical Caps
Yes
Polypropylene
120/Cs.
PCR Plate Cap Strip
96-well Plate