Thermo Scientific™ Domed PCR Caps, strips of 12
Domed cap strips for PCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.
Supplier:  Thermo Scientific™ AB0853
 View more versions of this product
Catalog No. AB-0853
Explore available promotions
Description
Description
Ensure proper sealing on all PCR strip tubes and 96-well PCR plates with precision molded Thermo Scientific™ PCR Plate Cap Strips.
Domed PCR Cap Strips are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Highlights:
  • Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
  • Large end tabs for strip removal (12 strips only)
Certifications:
Certified free from human DNA, RNases, and DNases.
Specifications
Specifications
Clear
12
Strips of 12 Domed Caps
Yes
Polypropylene
25 packs of 8
PCR Plate Cap Strip
96-well Plate