Learn More
Dopamine hydrochloride, 99%
CAS: 62-31-7 | C8H12ClNO2 | 189.64 g/mol
$80.15 - $923.26
Chemical Identifiers
| CAS | 62-31-7 |
|---|---|
| Molecular Formula | C8H12ClNO2 |
| Molecular Weight (g/mol) | 189.64 |
| MDL Number | MFCD00012898 |
| InChI Key | CTENFNNZBMHDDG-UHFFFAOYSA-N |
| Synonym | dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril |
| PubChem CID | 65340 |
| IUPAC Name | 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride |
| SMILES | [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1113606
|
Thermo Scientific Chemicals
A1113606 |
5 g |
N/A
|
|
|||||
|
AAA1113614
|
Thermo Scientific Chemicals
A1113614 |
25 g |
N/A
|
|
|||||
|
AAA1113622
|
Thermo Scientific Chemicals
A1113622 |
100 g |
N/A
|
|
|||||
Description
A neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA neurotransmitter and vasopressor that modulates cortical activation. Dopamine hydrochloride is a vasopressor that moderates cortical activation. It is used as a precursor to norepinepherine and epinephrine. It is an important neurotransmitter and chemical messenger that helps in the transmission of signals in the brain and other vital areas.
Solubility
Soluble in water, methanol and dimethyl sulfoxide. Insoluble in ether, petroleum ether, chloroform, benzene and toluene.
Notes
Store in cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 62-31-7 | |
| 189.64 | |
| CTENFNNZBMHDDG-UHFFFAOYSA-N | |
| 65340 | |
| [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1 |
| C8H12ClNO2 | |
| MFCD00012898 | |
| dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril | |
| 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride |
Specifications
| 62-31-7 | |
| 99% | |
| MFCD00012898 | |
| 3656720 | |
| 14,3421 | |
| Soluble in water,methanol and dimethyl sulfoxide. Insoluble in ether,petroleum ether,chloroform,benzene and toluene. | |
| [H+].[Cl-].NCCC1=CC=C(O)C(O)=C1 | |
| 189.64 | |
| 189.64 | |
| Dopamine hydrochloride |
| 243°C to 250°C | |
| C8H12ClNO2 | |
| 5 g | |
| Hygroscopic; Light sensitive | |
| dopamine hydrochloride, 3-hydroxytyramine hydrochloride, dopamine hcl, 4-2-aminoethyl benzene-1,2-diol hydrochloride, intropin, dopastat, revivan, dynatra, 3,4-dihydroxyphenethylamine hydrochloride, cardiosteril | |
| CTENFNNZBMHDDG-UHFFFAOYSA-N | |
| 4-(2-aminoethyl)benzene-1,2-diol;hydrochloride | |
| 65340 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 200-527-8
RTECSNumber : UX1092000
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only