missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Doppler Effect Demonstration Apparatus
Load batteries, swing the device in a full circular motion experience the doppler effect first hand. The Doppler Effect Demonstration Apparatus is the ideal demonstration of the Doppler effect in a classroom.
Supplier: Eisco™ DEF2013
Description
- The buzzer is 92 dB while standing still, but changes frequency when spun.
- Buzzer housing has a simple on/off switch and is constructed of rugged ABS plastic.
- Nylon cord is 2 ft. 8 in. attached to a rugged ABS handle 8.5 in. long, 1 in. dia.
- Demo comes with a 9 V battery installed.
Specifications
Doppler Effect Kit | |
9 V | |
9 V |
ABS Plastic | |
9 V Battery (Installed) |