missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific Chemicals Dorzolamide hydrochloride
$227.75 - $556.10
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC466610010
|
Thermo Scientific Chemicals
466610010 |
1 g |
Each for $556.10
|
|
|||||
AC466612500
|
Thermo Scientific Chemicals
466612500 |
250 mg |
Each for $227.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
130693-82-2 | |
C10H16N2O4S3·HCl |
Specifications
130693-82-2 | |
Solubility aq. soln.: soluble aq. soln.. Other solubilities: slightly soluble in methanol | |
C10H16N2O4S3·HCl | |
OSRUSFPMRGDLAG-CIRBGYJCSA-N | |
hydrogen (2R,4R)-4-(ethylamino)-2-methyl-1,1-dioxo-2H,3H,4H-1λâ¶-thieno[2,3-b]thiopyran-6-sulfonamide chloride | |
Dorzolamide hydrochloride |
1 g | |
281.0°C | |
360.89 | |
[H+].[Cl-].CCN[C@@H]1C[C@@H](C)S(=O)(=O)C2=C1C=C(S2)S(N)(=O)=O | |
360.89 |
Safety and Handling
GHS H Statement Harmful if swallowed.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
RUO – Research Use Only