United Scientific™ Double Convex Lens
Clear Lens for consistent optics demonstrations.
$4.03 - $9.65
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) Focal | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) Focal | Price | Quantity | ||||
S01324
|
United Scientific™
LCV304 |
38 mm | 100 mm |
Each for $4.03
|
|
||||
S01323
|
United Scientific™
LCV312 |
38 mm | 300 mm |
Each for $5.09
|
|
||||
S01322
|
United Scientific™
LCV508 |
50 mm | 200 mm |
Each for $4.03
|
|
||||
S01321
|
United Scientific™
LCV708 |
75 mm | 200 mm |
Each for $9.65
|
|
||||
Description
- Made from plate glass.
- Features carefully ground and polished optical surfaces.
- Cleanly ground edges.
- Double Convex.
Specifications
1 Ea. | |
Convex Lens |