Brady™ Double Hex Prolock
Effectively lock out a variety of isolation points. The Brady™ Double Hex Prolock will quickly become a staple lockout device in any facility. It tackles anything from T valve handles to electrical disconnects.
Supplier: Brady™ 122253
Description
- Designed to effectively lock out a variety of isolation points, including gate valves, T valve handles, electrical disconneccts, and more
- Economical and commact devices are highly visible and will quickly become a staple lockout device in any facility
Specifications
Pull-Tight Cable Lockout Device | |
Red | |
59 in. cable (red) |