missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Mopec Down Draft Work Cell
Mopec™ Down Draft Work Cell is a removable work surface that is designed to fit in a countertop and provides easy cleaning.
Supplier: Mopec BF600
Description
- Designed to fit in a 30 in. front to back countertop
- Removable work surface provides easy cleaning
- Down draft work cell has a 3.5 in. cup drain for any overflow and wash downs
- Using downdraft fume exhaust technology the BF600 down draft work cell provides a safe economical down draft working environment
- Ideal for working with reagents, formalin, xylene and alcohol
- Requires a sink base cabinet
Warranty
1 Year Warranty
Specifications
3.5 in. (8.89cm) Cup drain for any overflow | |
8 in. | |
20.32cm |
Down draft | |
68.58 in. |