missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™

Broad spectrum antibiotic

$191.00 - $402.45

Chemical Identifiers

CAS 10592-13-9
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.90
MDL Number MFCD03427564
InChI Key VLUQVUWDECWBTL-UQVCFKGQSA-N
PubChem CID 54706018
IUPAC Name [(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
SMILES [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP26531
SDS View Documents Promotion Details
Fisher BioReagents
BP26531
1 g Amber Glass
Each for $191.00
Case of 9 Each for $1,719.00
 
BP26535
SDS View Documents Promotion Details
Fisher BioReagents
BP26535
5 g Amber Glass
Each for $402.45
Case of 9 Each for $3,622.05
 
Description

Description

Broad spectrum antibiotic

Broad spectrum antibiotic. Broad spectrum inhibitor of matrix metalloproteinases in vivo.
Specifications

Chemical Identifiers

10592-13-9
480.90
VLUQVUWDECWBTL-UQVCFKGQSA-N
[(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
C22H25ClN2O8
MFCD03427564
54706018
[Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12

Specifications

Doxycycline Hydrochloride
10592-13-9
210°C
C22H25ClN2O8
Amber Glass
VLUQVUWDECWBTL-UQVCFKGQSA-N
[(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
54706018
Inclusive between 105°
Yellow
Doxycycline Hydrochloride
Powder/Solid
Inclusive between 300 at 349nm
95%
MFCD03427564
Inclusive between 800μg/mg
[Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12
480.90
480.9
Inclusive between 1.4%
1 g
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not induce vomiting. Call a physician or Poison Control Center immediately. . .

NFPA

Health:2
Flammability:0
Instability:0

Recommended Storage : 0° to 5°C, light sensitive, desiccate.

SDS
missing translation for 'documents'

missing translation for 'documents'