Dräger™ Clan Lab Simultest Reagent Set
Five-tube reagent set for use with Dräger Clan Lab Simultest Kit
Supplier: Dräger™ 8103310
Alert:
Avoid skin contact as contents are corrosive
Description
- Set includes ammonia, hydrochloric acid, iodine, phosgene and phosphine
Specifications
Clandestine Lab 5-tube Reagent Set | |
Ammonia, Hydrochloric Acid, Iodine, Phosgene, Phosphine tube |