Dräger™ CMS Chips: Hydrogen Cyanide
Suitable for 10 measurements of one gas
Supplier: Dräger™ 6406100
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifications
700 to 1100 hpa | |
1 to 20mg/L | |
30 to 260 sec. | |
0°C to 30°C |
Hydrogen Cyanide CMS Chip | |
2 to 50 | |
32°F to 104°F |