Dräger™ CMS Chips: Hydrogen Sulfide
Suitable for 10 measurements of one gas.
Supplier: Dräger™ 6406050
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.Specifications
700 to 1100 hpa | |
1 to 40mg/L | |
20 to 200 sec. | |
0°C to 40°C |
Hydrogen Sulphide CMS Chip | |
2 to 50 | |
32°F to 104°F |