Dräger™ CMS Chips: Hydrogen Sulfide
Suitable for 10 measurements of one gas.
Supplier:  Dräger™ 6406520
 View more versions of this product
Catalog No. 19-046641
Explore available promotions
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
40 to 450 sec.
0°C to 40°C
Hydrogen Sulphide CMS Chip
0.2 to 5
32°F to 104°F