Dräger™ CMS Chips: Chlorine
Suitable for 10 measurements of one gas
Supplier:  Dräger™ 6406010
Catalog No. 18-999-1922
Explore available promotions
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
5 to 12mg/L
30 to 400 sec.
0°C to 40°C
Chlorine CMS Chip
0.2 to 10
32°F to 104°F