Dräger™ CMS Chips: Hydrochloric Acid
Suitable for 10 measurements of one gas
Supplier:  Dräger™ 6406140
 View more versions of this product
Catalog No. 18-999-1924
Explore available promotions
Description
Description
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 10mg/L
6 to 80 sec.
0°C to 40°C
Hydrochloric Acid CMS Chip
20 to 500
32°F to 104°F